Thermo Scientific™ Finnpipette™ F1 Variable Volume Single-Channel Pipettes
Simplify volume adjustment with an easy “set and forget” lock mechanism, one of several convenient features of this ergonomic pipette.
Manufacturer: Thermo Scientific™ 4641140
 View more versions of this product
Catalog No. 14-386-293
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Model Finnpipette™ F1
    Description F1 Variable Volume Pipette
    Volume (Metric) Range 5 to 50μL
    Compatible Tips Flex 200, 300, 250 Univ, 200 Ext.,Flex 200, 300, 250 Univ, 200 Ext.
    Color Code Turquoise,Turquoise
    Volume Range (Metric) 5 to 50µL
    Simplify volume adjustment with Thermo Scientific™ Finnpipette™ F1 Variable-Volume Pipettes, which include an easy “set and forget” lock mechanism that features an adjustable finger rest and proprietary soft touch tip ejection. An antimicrobial treatment protects the instrument against contamination.
    Features:
    • Silver (Ag) ions create an antimicrobial surface that inhibits growth of bacteria, microbes, and fungus
    • “Set-and-forget” pipetting button enables easy volume adjustment
    • Pipetting button rotates freely when not in locking position
    • Finger rest can be adjusted 120° for optimal pipetting position
    • Light, smooth pipetting action enables better results over longer periods
    • Super blow-out piston with volumes of 50µL and below ensures delivery of micro-size drops
    • Proprietary soft-touch tip ejection minimizes forces needed for tip ejection
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback