Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes: PROMO with Trade in Available on GSA/VA Contract for Federal Government customers only.
Eliminate the possible effects of hand warmth on the accuracy of measurements with these pipettes, which feature a thermally-isolated AVG mechanism.
Manufacturer:  Thermo Scientific™ P4642130
 View more versions of this product
Catalog No. 14386295TI
Description
Description
  • Large Ergovisio display with white numbers on black background for increased visibility
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Light, smooth pipetting action enables better results over longer periods
  • Fully autoclavable
  • Super blow-out piston with volumes of 50µL and below ensures delivery of micro-size drops
Warranty: 5 years with web registration.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5 to 50μL
Finntip Flex 200, Finntip Flex 300, Finntip 250 universal, Finntip 200 Extended
0.1μL
Yellow
±0.3 to 0.15%
5 to 50μL
2.5 to 0.3%
Finpipette F2
Documents
Promotions
Promotions
  • Thermo Scientific
    Upgrade to a new Thermo Scientific Finnpipette F2 Manual Pipette and get $138 off the price of a single channel pipette or up to $277 off the price of a multichannel one.
    12-31-2019
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.