Learn More
Thermo Scientific™ Finnpipette™ Focus Fixed Volume Pipettes
Thermo Scientific™ Finnpipette™ Focus Short Fixed Volume Pipette has been discontinued, and is no longer available.
Manufacturer:  Thermo Scientific™ 4601040
 View more versions of this product
Catalog No. 14-387-02
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Thermo Scientific™ Finnpipette™ Focus™ Short Fixed Volume Pipette has been discontinued, and is no longer available. Please take a moment to learn about our current available manual pipetting systems.
Fixed-Volume Models
  • 1 to 25µL models feature “super blow-out” new streamlined design
  • Constructed of tough, lightweight polymers; fully autoclavable for absolute sterility
  • Easily disassembled for cleaning
Compatible with:
For use with Thermo Scientific Finntip tips. Also compatible with all Fisherbrand™ Universal-Fit tips, Fisherbrand SureOne, Fisherbrand Ergo-F, Large Orifice, Large Orifice/Genomic Aerosol Barrier, and Low-Retention tips.
Description & Specifications
Specifications
±0.6%
Finnpipette Focus Short Fixed, Volume: 20µL, Discontinued
0.5%
20µL
Accessories