Thermo Scientific™ Finnpipette™ Focus Fixed Volume Pipettes
Thermo Scientific™ Finnpipette™ Focus Short Fixed Volume Pipette has been discontinued, and is no longer available.
Manufacturer: Thermo Scientific™ 4601100
 View all options for this product
Catalog No. 14-387-08
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Accuracy ±0.3%
    Description Finnpipette Focus Short Fixed, Volume: 500µL, Discontinued
    Volume (Metric) 500µL
    Precision 0.3%
    Thermo Scientific™ Finnpipette™ Focus™ Short Fixed Volume Pipette has been discontinued, and is no longer available. Please take a moment to learn about our current available manual pipetting systems.
    Fixed-Volume Models
    • 1 to 25µL models feature “super blow-out” new streamlined design
    • Constructed of tough, lightweight polymers; fully autoclavable for absolute sterility
    • Easily disassembled for cleaning
    Compatible with:
    For use with Thermo Scientific Finntip tips. Also compatible with all Fisherbrand™ Universal-Fit tips, Fisherbrand SureOne, Fisherbrand Ergo-F, Large Orifice, Large Orifice/Genomic Aerosol Barrier, and Low-Retention tips.
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback