Learn More
Thermo Scientific™ Finnpipette™ Focus Short Variable Volume Pipettes
Thermo Scientific™ Finnpipette™ Focus Short has been discontinued, and is no longer available.
Manufacturer:  Thermo Scientific™ 4600110
 View more versions of this product
Catalog No. 14-387-17
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Specifications
Description
Finnpipette Focus Short has been discontinued, and is no longer available. Please take a moment to learn about our current available manual pipetting systems.
Features:
  • Digital display is easy to read even while pipetter is in use
  • “Super blow-out” feature on microsize pipetters ensures a new level of accuracy and precision at low-volume settings
  • Telescopic piston boosts the power of the blow-out step, overcoming the capillary action that can hinder delivery of residual liquid
  • Soft-touch tip ejection, an innovative gearing mechanism that produces increased downward force for tip release, also works to relieve hand stress
  • Calibration is simply performed using tool (provided)
Construction:
  • Constructed of tough, lightweight polymers; fully autoclavable for absolute sterility
  • Lightweight, ergonomic design defends against user fatigue
  • Pipetter disassembles easily for cleaning or parts replacement
  • Convenient stick-on shelf hanger allows you to store pipetter on a shelf, counter, or almost any surface with a square edge
  • Features “fast-dial” system with 0.01µL fine adjustment on microvolumes
  • Large, color-coded thumb button allows fast, accurate volume setting†
  • Finger rest ensures a secure grip and correct pipetting angle
† When a range is given, the first value applies to the smallest volume in the stated range; the second, to the largest volume.
Compatible with:
For use with Finntip™ tips. Also compatible with all Fisherbrand™ Universal-Fit tips, Ergo-F, Large Orifice, Large Orifice/Genomic Aerosol Barrier, and Low-Retention tips.
Includes:
Calibration certificate guarantees accuracy and precision; in-lab calibration tool.
Description & Specifications
Specifications
0.5 to 5µL (Micro)
Finntip 10
0.01µL
Pink
±3.5 to 1.0%
Finnpipette Focus Short µTip; Range: 0.5-5µL, Discontinued
3.0 to 0.8%
Accessories