Thermo Scientific™ Finnpipette™ Multistepper Pipette Available on GSA/VA Contract for Federal Government customers only.
Reduce the risk of error and increase productivity in immunoassay applications. Designed especially for work with microplates, featuring simultaneous 8-channel dispensing.
Manufacturer: Thermo Scientific™ 4540500
 View more versions of this product
Catalog No. 21-377-370
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description Handle and module
    Reduce the risk of error and increase productivity in immunoassay applications. The Thermo Scientific™ Finnpipette™ Multistepper Pipette is a lightweight, easy-to-use repeater pipette that provides simultaneous dispensing in eight channels. Designed especially for work with microplates, the Finnpipette Multistepper helps speed up routine laboratory procedures. Easy to use—simply attach, fill, and prime the module, then dial the selected volume.
    Design
    • Universal handle also fits the seven single-channel Thermo Scientific™ Finntip™ Steppers
    • Lightweight construction minimizes hand fatigue during repetitive dispensing
    • Separate tip ejector for safe and effortless tip disposal
    • Wide finger rest minimizes operator hand strain
    • Module rotates a full 360°, optimizing efficiency for both right or left-handed users
    Compatible with:
    Accepts Finntip Multistepper 1500µL tips. They are extra long, with more air space between the liquid and tip cone to prevent contamination.
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback