Thermo Scientific™ Finnpipette™ Stands Available on GSA/VA Contract for Federal Government customers only.
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands.
Manufacturer: Thermo Scientific™ 2209480
 View more versions of this product
Catalog No. 14-487-83
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Holds 3 Adapters for Carousel Stand
    Description Finnpipette Novus Adapter for Carousel Stand
    Compatibility 3 Adapters for Carousel Stand
    Item Description Finnpipette Novus Adapter for Carousel Stand, 3 Adapters
    Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands.
    Finnpipette F-Stand
    • Accomodates six pipettes
    • Sturdy crossbar structure with slot for each pipette for secure storage
    • Suitable for both single channel and multichannel models
    Finnpipette Multichannel Stand
    • Multi-position stand
    • Can hold any multichannel Finnpipette
    • Pipette can be stored in upright or horizontal position
    • Small footprint
    Finnpipette Carousel Stand
    • Rotating, portable
    • Transparent lid protects pipettes from contamination
    • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
    Finnpipette Novus Stand
    • For convenient storage of electronic Finnpipette Novus pipette
    • Accommodates both single and multichannel Finnpipette Novus models with ease
  • Description & Specifications
Related Products
Product Content Feedback