Thermo Scientific™ Finntip™ Biocon Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
Manufacturer: Thermo Scientific™ 94053200
 View more versions of this product
Catalog No. 21-377-359
  N/A
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Equipment) Finntip BioCon 250
    Volume (Metric) 250μL
    Product Line Finntip Biocon
    Filtered No
    Model Finntip BioCon 250
    Description Finntip BioCon 250, 0.5 to 250μL; 100/box
    Packaging 100/box, Sterile,100/box, Sterile
    Certifications/Compliance Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified).
    Sterility Sterile
    Perform PCR and other purity demanding applications with Thermo Scientific™ Finntip™ Biocon Pipette Tips, certified free from endotoxin, DNA, DNase and RNase.
    Features:
    • Individually packed
    • Three models in volume range 0.2-1000µL
    • Presterilized with gamma radiation
    • Includes certificate of quality
    Recommended for: PCR applications, molecular biology
  • Description & Specifications
Related Products
Product Content Feedback