Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finntip™ Flex Filter Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

Minimize the risk of cross-contamination. These tips combine the ergonomics of a standard pipette tip and the safety of a filter tip

Manufacturer:  Thermo Scientific™ 94056970

 View more versions of this product

Catalog No. 14-487-82


Add to cart

Description

Description

Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Thermo Scientific™ Finnpipette™
10mL
Sterile
Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified). Certificate of Conformity provided with each 10-rack case.
Finntip Flex
Finntip Flex Filter 10mL Ext., sterile
Filtered
26.7cm
Thermo Scientific™ Finnpipette™
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.