Learn More
Thermo Scientific™ Finntip™ Flex™ Pipette Specific Tips Available on GSA/VA Contract for Federal Government customers only.
Ensure a better seal with the pipetter module, for higher accuracy and precision.
Manufacturer:  Thermo Scientific™ 94060716
 View more versions of this product
Catalog No. 14-387-86
Add to cart
Description
Description
Ensure a better seal with the pipetter module, for higher accuracy and precision. The Thermo Scientific™ Finntip™ Flex™ Specific Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Thermo Scientific™ Finnpipette™
1000μL
Non-sterile
Finntip Flex 1000 refill starter kit
8.7cm
Thermo Scientific™ Finnpipette™
Refill
Non-Filtered
Blue
3.4 in.
ISO 9001, ISO 14001, ISO 13485
Finntip Flex
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Fulfills IVD Directive 98/79/EC requirements, valid only in Europe. Not for in vitro diagnostics use in North America.