Learn More
Thermo Scientific™ Finntip™ Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Ensure accurate, precise pipetting. Finntip™ Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
$109.00 - $1133.00
Specifications
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
Product Line Finntip
Filtered Non-filtered
View More Specs
Products 6
Catalog Number Mfr. No. Description Volume (Metric) Color Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) Color Sterility Price Quantity & Availability  
21-377-296 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400360
Finntip 50, Non-Filtered, Non-sterile, Bulk 50 μL Turquoise Non-sterile
Pack of 1000 for $109.00
Add to cart
21-377-385 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400370
Finntip 50, Non-Filtered, Non-sterile, Rack 50 μL Turquoise Non-sterile
Case of 3840 for $896.00
Add to cart
21-377-386 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400373
Finntip 50 Sterile, Non-Filtered, Sterile, Rack 50 μL Turquoise Sterile
Case of 3840 for $1,133.00
Add to cart
21-377-802 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400610
Finntip 20, Non-Filtered, Non-sterile, Rack 20 μL Purple Non-sterile
Case of 3840 for $640.00
Add to cart
21-377-801 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400613
Finntip 20 Sterile, Non-Filtered, Sterile, Rack 20 μL Purple Sterile
Case of 3840 for $810.00
Add to cart
21-377-800 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
9400620
Finntip 20, Non-Filtered, Non-sterile, Bulk 20 μL Purple Non-sterile
Pack of 1000 for $111.00
Add to cart
Description
Description
Ensure accurate, precise pipetting. Thermo Scientific™ Finntip™ Pipette Tips are manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips, to prevent liquid retention.
  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2µL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk, and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging
Note:
Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ISO 9001, ISO 14001, ISO 13485
Finntip
Non-filtered
SDS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.