Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIP200 Mouse anti-Human, Clone: N1-8, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | FIP200 |
---|---|
Clone | N1-8 |
Concentration | 0.5 mg/mL |
Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC174825
|
MilliporeSigma
MABC174825UL |
25 μL |
Each for $177.00
|
|
|||||
MABC1748100
|
MilliporeSigma
MABC1748100UL |
100 μL |
Each for $450.00
|
|
|||||
Description
FIP200 Monoclonal antibody specifically detects FIP200 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.Specifications
FIP200 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
N1-8 | |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
GST-tagged recombinant fragment corresponding to 247 amino acids from the N-terminal region of human FIP200 and a C-terminal polypeptide fragment of FIP200. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title