Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fir Needle Oil, Siberian Type, FCC, 32-44%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
8021-29-2
8021-29-2

F1078, 8021-29-2

Spectrum™ Fir Needle Oil, Siberian Type, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Specific Gravity 0.898-0.912
Refractive Index 1.468-1.473
Quantity 100 mL
CAS Max % 1
UN Number UN1272
Percent Purity 32 to 44%