Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fir Needle Oil, Siberian Type, FCC, 32-44%, Spectrum™ Chemical
SDP

Catalog No. 18602733 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-733 500 mL Amber Glass Bottle
18-602-732 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-733 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1078500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F1078, 8021-29-2

Spectrum™ Chemical Fir Needle Oil, Siberian Type, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8021-29-2

Specifications

CAS Max % 100%
Boiling Point 132 °C (270 °F) - lit.
Packaging Amber Glass Bottle
Refractive Index 1.468-1.473
Quantity 500 mL
UN Number UN1272
Specific Gravity 0.898-0.912
Percent Purity 32 to 44%
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Fir Needle Oil, Siberian Type
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.