Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIR Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5104510

Catalog No. PIPA5104510


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total FIR.

FARP2 (FERM, RhoGEF and pleckstrin domain-containing protein 2), also known as PLEKHC3 or FERM domain including RhoGEF (FIR), is a 1,545 amino acid protein that contains one FERM domain, one DH domain and two PH domains. It exists as two alternatively spliced isoforms that are abundantly expressed in brain, lung, and testis as well as in embryonic hippocampal and cortical neurons. FARP2 functions as a Rho-guanine nucleotide exchange factor that activates RAC1 and is thought to regulate neurite remodeling of embryonic neurons. Sema3A binding to neuropilin-1 induces the dissociation of FARP2 from plexin-A1, thereby activating FARP2's Rac GEF activity which is critical for repulsion of outgrowing axons and suppression of neuronal adhesion. Downregulation of the FARP2 gene has been implicated in autism.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FIR
Polyclonal
Unconjugated
FARP2
AI465173; BC009153; D030026M03Rik; Farp2; FERM domain including RhoGEF; FERM domain-including RhoGEF; FERM, ARH/RhoGEF and pleckstrin domain protein 2; FERM, ARHGEF and pleckstrin domain-containing protein 2; FERM, RhoGEF and pleckstrin domain protein 2; FERM, RhoGEF and pleckstrin domain-containing protein 2; FGD1-related Cdc42-GEF; Fir; FRG; Kiaa0793; mKIAA0793; PH domain-containing family C member 3; pleckstrin homology domain-containing family C member 3; PLEKHC3
Rabbit
Affinity chromatography
RUO
227377, 9855
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O94887, Q91VS8
FARP2
A synthesized peptide derived from human FARP2(Accession O94887), corresponding to amino acid residues S344-L394.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.