Fire Hooks Unlimited Heavy Duty Contractor Bolt Cutters
For different cutting tasks including locks, chain links, reinforcing rods, bolts, rivets
Manufacturer: Fire Hooks Unlimited IncSupplier Diversity Partner BC24"
 View more versions of this product
Catalog No. 19-811-457
  Check Availability
Add to Cart
    Specifications
    Product Type Bolt Cutters
    For Use With (Application) Cutting bolts up to 3/8 in. dia.
    Length (English) 24 in.
    • Available in different sizes: 18 in., 24 in.
  • Description & Specifications
Related Products
Product Content Feedback