Fire Hooks Unlimited K-Tools
Designed (by a FDNY Fire Lieutenant & licensed locksmith) to pull all types of lock cylinders (rim or mortise) from various glass type or solid doors
Manufacturer: Fire Hooks Unlimited IncSupplier Diversity Partner KTOOL
 View more versions of this product
Catalog No. 19-811-473
  Check Availability
Add to Cart
    Specifications
    Product Type K-Tool
    For Use With (Application) Pull lock cylinders of virtually any type including mortise, tubular and rim
    Includes K-Tool, instructions
  • Description & Specifications
Related Products
Product Content Feedback