Learn More
BioworldSupplier Diversity Partner FireTag™ HRP Conjugation Kit, 1 Kit DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220800001
FireTag-HRP Conjugation Kit Based on well-established periodate chemistry, yielding consistent and reproducible protein conjugates. This kit contains all reagents required to conjugate your protein or antibody, including buffers and activated HRP and requires no activation or purifi cation. Suffi cient reagents are provided to conjugate 6 samples, each containing up to 0.1 mg of antibody or protein. Modifi ed HRP, SFB Linker and AP Conjugation Buffer (5X). FireTag-Activated HRP is also available as a stand-alone reagent. See catalog # 600048
Catalog No. 50-253-998