Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

First Safety™ Gas Cylinder Brackets

All-steel construction wall mounted brackets

Manufacturer:  FIRST SAFETY CORPORATION G100

 View more versions of this product

Catalog No. 17-987-155A


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

NFPA1, CalOSHA, UFC
Gas cylinder
2 in.
One cylinder in the vertical position from 4 to 12 in dia.
8 in.
Steel
Bracket
1.36kg
10.16cm
8 x 4 x 2 in. (20.3 x 10.16 x 5cm)
11g
5cm
Woven polypropylene straps, steel cinch style buckles
20.3cm
Wall
3 lb.
4 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit