Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOWDiagnostics, Inc. First To Know Rapid Syphilis Test
First To Know Rapid Syphilis test is the first FDA-cleared point-of-care test for self-collection and over-the-counter detection of Syphilis. This CLIA-waived test offers a quick and accurate result, bringing you a convenient new syphilis testing option.
$479.00 - $860.00
Specifications
Clia Complexity | Waived |
---|---|
Detectable Analytes | Syphilis RPR Antigen |
Type | Syphilis Test |
Disposable | Single-use |
Form | Ready-to-Use (RTU) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
21-377-374
|
NOWDiagnostics, Inc.
600736 |
1 Test |
Case of 12 for $479.00
|
|
|||||
21-377-375
|
NOWDiagnostics, Inc.
600737 |
25 Test |
Case of 25 for $860.00
|
|
|||||
Description
- FDA cleared, over-the-counter, CLIA waived, 15 minute result
- Able to be performed by any users, qualitative result (positive = test line)
- Only cleared product that has fingerstick and blood drop directly on the device, no separation or sample prep needed - just Touch, Tap, and Test.
- Intended for symptomatic or asymptomatic patients with no prior history of syphilis infectionThe First To
- Know Rapid Syphilis test is the only FDA cleared POCT device for self collect and OTC
Specifications
Waived | |
Syphilis Test | |
Ready-to-Use (RTU) |
Syphilis RPR Antigen | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title