Learn More
fischertechnik™ Education Physics I Model Set Products for Science Education
To learn about physical phenomena such as acceleration, inertia of mass, laws of mass, and more. fischertechnik™ Education Physics I Set includes track based, coaster style models to make physics engaging and easy to understand.
Manufacturer:  fischertechnik™ 533050
Catalog No. S35684
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Not for children under five--product contains small parts which can be swallowed * Electronic parts contain lead. May be harmful if eaten or chewed. May generate dust containing lead.
Description
Description
 690 parts for building 7 different models
Warranty and Services
2 years from date of purchase
Specifications
Specifications
Recommended Students Per Set: 2
Full color assembly manual, Multi language teaching and activity book
Accessories/Batteries: Accu Set or Power Set, or 9v battery
Middle school, high school, and college STEM education, principles of physics
Physics I Model