Learn More
fischertechnik™ Education Electronics Set Products for Science Education
To study basic principles of electronics. fischertechnik™ Electronics Set helps students to build functional models from simple flash light to ship see-saw, alternating flasher and controllable ventilating fan.
Manufacturer:  fischertechnik™ 533029
Catalog No. S35682
Add to cart
Alert:
Not for children under five--product contains small parts which can be swallowed * Electronic parts contain lead. May be harmful if eaten or chewed. May generate dust containing lead.
Description
Description
  • 260 parts for building 16 instructional models
Following models can be built: Simple Electrical Circuit, Flashlight, Continuity Tester, Refrigerator Light, Stamping Machine, Parallel Connection, Hallway Lighting, Merry-Go-Round with Pole Reversing Switch, Simple Flasher, Alternating Flasher, Touch Switch Merry-Go-Round, Merry-Go-Round with Light Barrier, Merry-Go-Round with Touch Switch, Ship See-Saw, Ship See-Saw with Alternating Flasher, Alarm System, Ventilating Fan, Bathroom Fan, Sliding Door
Specifications
Specifications
Recommended Students Per Set: 2
Complete instructions along with multi-language teaching and activity booklet, Electronic-module, XS motor, 2x transistor, 2x capacitor, 3x resistor, 2x mini-switch, phototransistor, temperature sensor, lens tip lamp, 2x LED, battery tray for 9V block.
Electronics Set
9V battery