Learn More
Molecular Probes™ FISH Tag™ DNA Multicolor Kit, Alexa Fluor™ dye combination Non-distribution product as customer accommodation.
FISH Tag™ DNA Multicolor Kit, Alexa Fluor™ dye combination
Manufacturer:  Molecular Probes™ F32951
Catalog No. F32951
Add to cart
Description
Description
The FISH Tag™ DNA Multicolor Kit provides a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. The FISH Tag™ DNA Multicolor Kit contains fluorescent dyes in four colors, making it ideal for simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content. We also offer FISH Tag™ Kits in single colors.
FISH Tag™ Kit Specifications:
  • Dye (Ex/Em): Alexa Fluor™ 488 (492/520 nm), Alexa Fluor™ 555 (555/565 nm), Alexa Fluor™ 594 (590/615 nm), Alexa Fluor™ 647 (650/670 nm)
  • Comprehensive kit includes 16 reagents
  • Comes with detailed protocol to help ensure your success
    Complete Workflow Solution for FISH
    FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.
    More Options for Nucleic Acid Labeling
    To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.
    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
  • Specifications
    Specifications
    Fluorescent
    Store in freezer (-5 to -30°C) and protect from light.
    DNA
    Alexa Fluor™, FISH Tag™
    Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, Alexa Fluor™ 647
    Dry Ice
    Not Target-Specific
    SDS
    Documents
    Provide Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    For Research Use Only. Not for use in diagnostic procedures.