Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molecular Probes™ FISH Tag™ RNA Multicolor Kit, Alexa Fluor™ dye combination
FISH Tag™ RNA Multicolor Kit, Alexa Fluor™ dye combination
Supplier: Molecular Probes™ F32956
Description
The FISH Tag™ RNA Multicolor Kit provides a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. The FISH Tag™ RNA Multicolor Kit contains fluorescent dyes in four colors, making it ideal for the simultaneous localization of multiple sequence-specific RNA species in cells, tissues, and whole embryo mounts. We also offer FISH Tag™ Kits in single colors.FISH Tag™ Kit Specifications:
Complete Workflow Solution for FISH
FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.
More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook. For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Kit | |
Alexa Fluor™, FISH Tag™ | |
RNA | |
Dry Ice |
Fluorescence | |
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647 | |
DNA In Situ Hybridization Assay Kit | |
Not Target-Specific |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction