Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Fish Triiodothyronine ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of fish triiodothyronine.
Supplier: Novus Biologicals™ NBP2601821KIT
Description
The Fish Triiodothyronine ELISA Kit (Colorimetric) measures fish Triiodothyronine in serum, plasma and tissue homogenates.Specifications
0.5to 8ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of fish T3 | |
ELISA | |
1 Kit | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Triiodothyronine |
0.5ng/mL | |
Plasma, Serum, Tissue Homogenates | |
Fish | |
Assay plate, Conjugate, HRP-avidin, Wash Buffer (20 x concentrate), Substrate B, Substrate A, Standard, Stop Solution, Instruction manual | |
RUO | |
50 to 100 μL | |
liothyronine; T3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction