Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ 2M Calcium Chloride, Microbial Cell Culture Grade
Shop All Fisherbrand Products
Click to view available options
:
5 x 10mL
Description
- CAS: 10035-04-08 (Calcium Chloride Dihydrate), 7732-18-5 (Water)
- Concentration: 294g/L
- Calcium chloride solution is commonly used in cell biology to prepare competent E. coli cells with improved transformation efficiency
- Calcium chloride increases the ability of the cell to incorporate plasmid DNA due to the postitively charged calcium ions
- Promotes the binding of plasmid DNA to lipopolysaccharides
- Product is supplied as 5 x 10mL
- Intended for research use only

Specifications
Specifications
Concentration | 294g/L |
Form | Liquid |
Grade | Microbial cell culture |
Manufacturing Origin | United States |
Fill Volume | 50 mL |
For Use With (Application) | Widely used to prepare competent Escherichia coli strains |
Packaging Type | Bottle |
Quantity | 50 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction