Fisher BioReagents 2M Magnesium Chloride, Microbial Cell Culture Grade
Media, Prepared; 2M Magnesium Chloride; Fisher BioReagents; Microbial Cell Culture Grade; For addition to SOB and SOC medium for propagating bacterial strains; 406.6g/L; CAS: 7791-18-6 (MgCl2.6H2O), 7732-18-5 (Water)
Catalog No. BP9741-10X5
  Check Availability
Add to Cart
Custom packing/Bulk ordering
You can now order this item in custom quantities.  Learn More
  • Description & Specifications
  • Special Offers
Related Products
Product Content Feedback