Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Bovine Serum Albumin Solution Available on GSA/VA Contract for Federal Government customers only.

Specifications

Concentration 20mg/mL
Format Solution
For Use With (Application) Protein Stabilizer and Blocking Agent
Name BSA Solution
Source Bovine
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP675-1 Available on GSA/VA Contract for Federal Government customers only.
View Documents
1mL
Each for $119.00
Order in Bulk

Request bulk or custom formats

N/A
Due to product restrictions, please to purchase or view availability for this product.
BP675-5
View Documents
5mL
Each for $425.00
Order in Bulk

Request bulk or custom formats

N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

CAS: 9048-46-8

Bovine Serum Albumin is used as a protein stabilizer and blocking agent.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

20mg/mL
Protein Stabilizer and Blocking Agent
Bovine
Solution
BSA Solution
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit