Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ DNA Polymerase I Large (Klenow) Fragment (Recombinant E. coli Strain)

Specifications

Concentration 5 to 10U/μL
Product Type DNA Polymerase I
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP3201-1
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
BP32015
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Description: The DNA Polymerase I Large (Klenow) Fragment lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I but retains the 5' to 3' polymerase, the 3' to 5' exonuclease and strand displacement activities
  • Application: DNA Polymerase I Large (Klenow) Fragment is used for filling-in or labeling 3' ends of double-stranded DNA, second strand cDNA synthesis, and DNA sequencing by the dideoxy method
  • Storage Buffer Components: 0.05M Tris-HCl (pH 7.5), 1mM DTT, 0.1mM EDTA and 50% (v/v) glycerol
  • Provided with 10X Reaction Buffer: 500mM Tris-HCl (pH 7.2), 100mM MgSO4, and 1mM DTT
  • Unit Definition: One unit is defined as the amount of enzyme required to catalyze the incorporation of 10nmol of total deoxyribonucleotides into TCA-insoluble form in 30 minutes at 37°C in 67mM potassium phosphate, (pH 7.5), 6.7mM MgCl2, 1mM DTT, 133µM activated calf thymus DNA and 33µM dATP, dCTP, dGTP, and radiolabeled dTTP
  • Storage: -20°C

Specifications

Specifications

5 to 10U/μL
DNA Polymerase I
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit