Learn More
Fisher BioReagents™ exACTGene™ Core Reagent Set A
Optimized for routine PCR applications
 View more versions of this product
Catalog No. FB6210
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Provide an economical alternative for high quality PCR
  • Kit components are also available separately
Specifications
Specifications
Taq DNA Polymerase, 10X Buffer A, dNTPs
80.0
Core Reagent Set A
-20°C