Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents Taq DNA Polymerase
Allows up to 100 cycles of amplification without loss of activity
Description
- Licensed for use in PCR
- Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
- Has a 5'
3' exonuclease activity
- Available with two different reaction buffers
- Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
- Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
- Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween 20, 0.5% Nonidet P40
- Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial
Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above
Specifications
5U/μL | |
Liquid | |
Poly Tube | |
-20°C | |
500 Units |
PCR | |
10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial | |
Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5% | |
Taq DNA Polymerase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title