Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Taq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.

Allows up to 100 cycles of amplification without loss of activity

 View more versions of this product

Catalog No. FB-6000-70


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

  • Licensed for use in PCR
  • Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
  • Has a 5'3' exonuclease activity
  • Available with two different reaction buffers
  • Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
  • Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
  • Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
  • Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial

Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

5U/μL
Liquid
Poly Tube
-20°C
2500 Units
PCR
10X assay buffer B : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 25mM solution in a separate vial
Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5%
Taq DNA Polymerase
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit