Learn More
Fisher BioReagents™ M-MLV Reverse Transcriptase
Ideal for dependable first-strand cDNA synthesis
Specifications
Packaging Poly Tube
View More Specs
Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP3208-1
View Documents
10,000 Units This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP32085
View Documents
50,000 Units This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • >99% triphosphate
  • Chemically synthesized to meet a high purity standard
  • Ensure consistent results and optimum sensitivity for many rigorous applications
  • Tested for purity by HPLC, performance in PCR with 500 bp and 6kb templates, and absence of DNase, RNase, and protease activity to ensure dependable performance and lot-to-lot consistency
Specifications
Specifications
Poly Tube