Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Modified Terrific Broth (Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
$286.50 - $2410.00
Specifications
Format | Capsule |
---|---|
Media Type | Terrific Broth |
Product Type | Microbiology Media |
Formulation | Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
Catalog Number | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Packaging Type | Price | Quantity & Availability | ||||||
BP9729-600
|
Poly bottle, 25 Capsules (Each capsule contains 24g of broth) |
Each for $286.50
Order in Bulk
Request bulk or custom formats |
|
||||||
BP97295
|
Poly pail, 25 Capsules (Each capsule contains 24g of broth) |
Each for $2,410.00
Order in Bulk
Request bulk or custom formats |
|
||||||
Description
- Directions for Media Preparation: Add 2 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Greater peptone and yeast concentration allow for higher plasmid yield
- Product is in capsule form
- Each capsule contains 24g of broth
- Format: Capsule
Specifications
Capsule | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures |
Terrific Broth | |
Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title