Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Nucleotides

Nucleotides for PCR with a purity level of >98% triphosphate

Specifications

Format Liquid
pH 7 to 7.5
Concentration 100 mM
Purity >98%
Color Colorless
View More Specs

Products 2
Catalog Number Product Type Quantity Price Quantity & Availability  
Catalog Number Product Type Quantity Price Quantity & Availability  
BP25641
View Documents
dNTP Set 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product
N/A
BP25644
View Documents
dNTP Set 400 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Functionally tested in PCR
  • Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
  • DNase-, RNase-, and protease-free
  • PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
  • dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)

Specifications

Specifications

Liquid
100 mM
Colorless
7 to 7.5
>98%
Dideoxy sequencing, mutagenesis and cDNA synthesis
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit