Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ OptiGrow™ LB Broth Lennox Microbiological Media
Classic media in ready-to-use liquid form
Description
- Sterilized by autoclaving
- For high yield of bacterial culture
- No dusty powders to cause environmental concerns
- Fully automatic manufacturing process eliminates unexpected contamination
- LB Broth. Standard bacteriological nutrient medium for the propagation of E. coli for purposes of strain maintenance (sub-culturing), cloning, plasmid propagation, and protein expression
- Super Broth. Specifically developed to increase the yield of high copy number plasmids derived from suspension cultures of E. coli strains
- Terrific Broth. Supports higher cell densities than LB medium, increases plasmid DNA yield
Order Info
Supplied in case of ten 1L bottles
Specifications
Yellow | |
United States | |
10 x 1000 mL | |
10 x 1 L |
Liquid | |
LB Broth, Lennox | |
Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction