Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents Recombinant DNase 10X RXN Buffer, Optizym, Fisher BioReagents
Ideal for use in RT-PCR and other RNA applications
Specifications
Product Type | OPTIZYME™ rDNase I |
---|---|
Format | Viscous Liquid |
For Use With (Application) | RT-PCR, other RNA applications |
Storage Requirements | -20°C |
Catalog Number | Concentration | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Concentration | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
BP32261
|
2U/μL | 1000U | 20mM HEPES (7.5 pH), 10mM CaCl2, 10mM MgCl2, 1mM DTT, 50% Glycerol |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
BP32271
|
10X | Each | 100mM Tris HCl (7.5 pH), 25mM MgCl2, 5mM CaCl2 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
BP32262
|
2U/μL | 2,000U | 20mM HEPES (7.5 pH), 10mM CaCl2, 10mM MgCl2, 1mM DTT, 50% Glycerol |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Efficiently hydrolyzes phosphodiester linkages of double- or single-stranded DNA
- Requires magnesium and calcium for maximal activity
- Tested for RNase activity, protease activity, and in specific performance tests
- Storage Buffer Components: 20mM HEPES (pH 7.5), 10mM CaCl2, 10mM MgCl2, 1mM DTT, 50% (v/v) glycerol
- Provided with 10X Reaction Buffer: 100mM Tris-HCl (pH 7.5), 25mM MgCl2, 5mM CaCl2
- Unit Definition: One unit is defined as the amount of enzyme required to completely degrade 1µg DNA in 10 minutes at 37°C
- Unit present in each tube: 1mL per ampule
Specifications
OPTIZYME™ rDNase I | |
Viscous Liquid | |
RT-PCR, other RNA applications | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title