Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents Optizyme Ribonuclease Inhibitor, Recombinant
Protects RNA from degradation by RNases
$199.00
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
BP3222-1
|
2,500U |
Each for $199.00
Order in Bulk
Request bulk or custom formats |
|
BP32225
|
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Supplied as a recombinant product from E. coli (originally isolated from rat lung) and provides superior protection of RNA from degradation by RNases
- Commonly added to all solutions used during the isolation of RNA, and will not interfere with the performance of most enzymes in downstream applications
- Active over a broad temperature range, and even provides some RNase inhibition at 60°C, which is useful when performing reverse transcription reactions at elevated temperatures to overcome secondary structure in RNA
- Storage Buffer Components: 20mM HEPES-KOH (pH 7.6 at 4°C), 50mM KCl, 8mM DTT, and 50% glycerol (v/v)
Inactivates RNase A, RNase B, and RNase T2; No activity against SP6, T7, or T3 RNA polymerases, AMV or MMLV reverse transcriptases, and Taq DNA polymerase
- Displays a broad spectrum of inhibitory activity against RNases, and does not have activity against other polymerases and reverse transcriptases
- Suitable for use in common molecular biology applications such as isolation and purification of RNA, cDNA synthesis, RT-PCR, in vitro RNA transcription/translation, ribonuclease protection assay, and preparation of RNase-free antibodies
Specifications
RNase Inhibitor |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title