Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Restriction Enzymes - Hind III (Source: Haemophilus influenzae Rd)

Specifications

Concentration 10U/μL
Cut Site A.AGCTT
Product Type HindIII
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP33721
View Documents
5,000U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP33725
View Documents
15,000U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Application: Hind III restriction endonuclease digests dsDNA with sequence indicated below
  • Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 250mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
  • Qualified for Blue/White Cloning Assays
  • Storage: -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Specifications

10U/μL
A.AGCTT
HindIII
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit