Learn More
Fisher BioReagents™ Restriction Enzymes - Msp I (Source: Moraxella species)
 View more versions of this product
Catalog No. BP33845
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
·    Application: Msp I restriction endonuclease digests dsDNA with sequence indicated below
·    Storage Buffer Components: 10mM Tris-HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA and 50% glycerol
·    Storage: -20°C
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
MspI
10U/μL
C.CGG
10,000U