Learn More
Fisher BioReagents™ SOC Medium
Catalog No. BP9740-10X5
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Nutrient-rich growth media used to maximize the transformation efficiency of competent E. coli cells
  • Liquid media is conveniently packaged and supplied as 5 x 10mL bottles
Specifications
Specifications
Casein Peptone 20g/L, Yeast extract, 5g/L, Sodium Chloride 0.58g/L; Potassium Chloride 0.19g/L, Magnesium Chloride 0.95g/L, Magnesium Sulfate 1.20g/L, Glucose 3.60g/L
United States
Liquid
Bottle
10 mL
5 x 10mL
SOC Medium