Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ T4 DNA Polymerase (Source: Recombinant E. coli Strain)

Specifications

Concentration 5 to 10U/μL
Product Type T4 DNA Polymerase
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP32025
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP3202-1
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Application: T4 DNA Polymerase is used to flush 5' protruding ends with labeled or unlabeled dNTPs and blunt 3' overhangs. Also used in in vitro mutagenesis.
  • Description: T4 DNA Polymerase catalyzes the 5' to 3' synthesis of DNA from a primed single-stranded DNA template. Although possessing a potent 3' to 5' proofreading exonuclease, T4 DNA Polymerase contains no 5' to 3' exonuclease activity.
  • Storage Buffer Components: 200mM potassium phosphate (pH 6.5), 2mM DTT and 50% glycerol.
  • Unit Definition: One unit is defined as the amount of enzyme required to catalyze the incorporation of 10nmol of total nucleotide into an acid-insoluble form in 30 minutes at 37°C.
  • Storage: -20°C

Specifications

Specifications

5 to 10U/μL
T4 DNA Polymerase
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit