Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Terminal Deoxynucleotidyl Transferase (Recombinant)

Specifications

Concentration 15 to 30U/μL
Product Type Terminal Deoxynucleotidyl Transferase
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP3203-1
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP32035
View Documents
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Applications:

Application: Terminal Deoxynucleotidyl Transferase is used in tailing reactions to add complementary homopolymer tails to DNA vectors and cDNA and 3' end-labeling.

Specifications

Specifications

15 to 30U/μL
Terminal Deoxynucleotidyl Transferase
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit