Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Yeast Lytic Enzyme (From Arthrobacter luteus, 5000U/g)

Specifications

Concentration 5,000U/g
Product Type Yeast Lytic Enzyme
View More Specs

Products 1
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
BP268325
View Documents
N/A 25g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Application: This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with beta-1,3-linkages. It is useful for preparing yeast spheroplasts.
  • Lytic Activity: 5000U/g
  • Unit Definition: One unit will produce a ΔA800 of 0.001 per minute at pH 7.5 and 25°C using a suspension of brewers yeast as substrate in a 3mL reaction mixture.
  • Storage: 0° to 5°C
Specifications

Specifications

5,000U/g
Yeast Lytic Enzyme
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit