Available on GSA/VA Contract for Federal Government customers only. Fisher Diagnostics™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (aPTT) Reagents

aPTT reagents provide sensitivity to intrinsic and common pathway factors, heparin and lupus anticoagulant

Pacific Hemostasis Activated Partial Thromboplastin Time reagents (APTT) are used to identify acquired or congenital disorders to the intrinsic and common pathways. APTT testing is widely accepted as a means of monitoring patients on oral anticoagulant therapy such as heparin.

Specifications

Clia Complexity Moderate
View More Specs
  • Products 7
  • Description & Specifications
Products
Catalog Number Product Type Quantity Price Quantity & Availability    

022924

KONTACT (Micronized Silica Activator) 10 x 10mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

285545

Available on GSA/VA Contract for Federal Government customers only.  SDS
CaCl2, 0.02M 10 x 10mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

022925

SDS
CaCl2, 0.025M 10 x 4mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

022927

Available on GSA/VA Contract for Federal Government customers only. 
aPTT-XL 10 x 10mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

176149

SDS
CaCl2, 0.025M 10 x 10mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

022929

APTT-XL (Ellagic Acid Activator) 10 x 4mL N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 

022926

SDS
aPTT-LS 10 x 10mL N/A N/A N/A
 This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
Descriptions & Specifications

Related Products