Fisher Diagnostics™ PROTOCOL™ Individual Gram Stains Available on GSA/VA Contract for Federal Government customers only.

Manufactured with traditional formulations used by most laboratories

 View more versions of this product

Catalog No. 2300584

  Check Availability
Add to Cart

    Specifications

    Composition Water, Polyiodine complex, Potassium Iodide
    Stain Type Gram Stain
    Type Iodine
    Quantity 8 oz.
    Includes Dispenser cap
    Format Bottle
    Color Dark Red, Dark Brown

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit