Fisher HealthCare™ Accessories for Tissue Flotation Baths and Flotation Work Stations: Slide Drying Racks
For drain-drying slides
 View more versions of this product
Catalog No. 22-047-781
  Check Availability
Add to Cart
    Specifications
    Product Type Slide drying rack
    For Use With (Equipment) 8 x 8 in. and 8 x 11 in. Model tissue flotation baths and flotation work station
    Includes 13 x 1 in. trough for blotting paper
    Mounting Type Side Mounted
    Holds 34 slides
  • Description & Specifications
Related Products
Product Content Feedback