Fisher HealthCare™ Accessories for Tissue Flotation Baths and Flotation Work Stations: Slide Drying Racks
For drain-drying slides
 View more versions of this product
Catalog No. 22-047-779
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 8 x 11 in. Model tissue flotation baths and flotation work station
    Holds 39 slides
    Product Type Slide drying rack
    Includes 13 x 1 in. trough for blotting paper
    Mounting Type Front Mounted
  • Description & Specifications
Related Products
Product Content Feedback