Fisher HealthCare™ PROTOCOL™ Color-Coded Prefilled Formalin Containers Available on GSA/VA Contract for Federal Government customers only.

Prefilled with 10% neutral buffered formalin

 View more versions of this product

Catalog No. 23-316155

  Check Availability
Add to Cart

    Specifications

    Volume (Metric) 30mL formalin fill
    Label Paper
    Closure Color Blue
    Graduated Yes
    Material Autoclavable Polypropylene
    Cap Type Color-coded cap w/leakproof O-ring
    Capacity (Metric) 60mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit