Learn More
Fisher HealthCare™ PROTOCOL™ Empty Containers Available on GSA/VA Contract for Federal Government customers only.
Eliminate formalin evaporation
$217.50 - $718.00
Specifications
Label Adhesive
Material Polypropylene
View More Specs
Products 5
Catalog Number Capacity Includes Price Quantity & Availability  
Catalog Number Capacity Includes Price Quantity & Availability  
22-032061 Available on GSA/VA Contract for Federal Government customers only.
View Documents
0.68 oz.; 20mL 200 Containers
Case of 200 for $239.00
Add to cart
22-032062 Available on GSA/VA Contract for Federal Government customers only.
View Documents
4.0 oz., 120mL 300 Containers
Case of 300 for $718.00
Add to cart
22-316158 Available on GSA/VA Contract for Federal Government customers only.
View Documents
1.375 oz., 40mL 600 Containers
Case of 600 for $311.50
Add to cart
22-316160
View Documents
3.04 oz.; 90mL 400 Containers
Case of 400 for $217.50
Add to cart
22-316159 Available on GSA/VA Contract for Federal Government customers only.
View Documents
2.0 oz., 60mL 500 Containers
Case of 500 for $529.00
Add to cart
Description
Description
  • Polypropylene
  • Autoclavable
  • Graduation marks clearly identify volume
  • Empty containers come with adhesive labels for patient identification and hazardous chemical warning
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Adhesive
Polypropylene